Assay Details
| Gene Symbol: TNNI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003281.3 | NP_003272.3 | 6 - 7 | 373 | 81 | |
| GenBank mRNA | AJ315823.1 | - | 6 - 7 | 353 | 81 | |
| AK223588.1 | - | 6 - 7 | 355 | 81 | ||
| AK311896.1 | - | 6 - 7 | 357 | 81 | ||
| AL831820.1 | - | 6 - 7 | 357 | 81 | ||
| AL831975.1 | - | 6 - 7 | 357 | 81 | ||
| BC012600.1 | - | 6 - 7 | 361 | 4245163 | 81 | |
| BC012601.1 | - | 6 - 7 | 352 | 4245718 | 81 | |
| BI825503.1 | - | 5 - 6 | 373 | 81 | ||
| CR450301.1 | - | 3 - 4 | 280 | 81 | ||
| CR541996.1 | - | 3 - 4 | 280 | 81 | ||
| CR542030.1 | - | 3 - 4 | 280 | 81 | ||
| J04760.1 | - | 6 - 7 | 356 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003689.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01056664.hg.1 | ENST00000336092.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056664.hg.1 | ENST00000358712.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056664.hg.1 | ENST00000361379.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056664.hg.1 | ENST00000367312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056664.hg.1 | ENST00000413495.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056664.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056664.hg.1 | TC01003689.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000555948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000555948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000555948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000555948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000555948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000555948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056663.hg.1 | ENST00000336092.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056663.hg.1 | ENST00000358712.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056663.hg.1 | ENST00000361379.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056663.hg.1 | ENST00000367312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056663.hg.1 | ENST00000413495.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056663.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056663.hg.1 | TC01003689.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000555948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000555948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000555948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000555948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000555948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000555948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016914.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228086.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228086.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228086.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228086.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228086.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228086.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228086.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228086.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228086.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228086.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228086.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228086.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228086.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228086.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228086.hg.1 | TC0100016914.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100228087.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228087.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228087.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228087.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228087.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228087.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228087.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228087.hg.1 | ENST00000336092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228087.hg.1 | ENST00000361379.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228087.hg.1 | ENST00000367312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228087.hg.1 | ENST00000555340.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228087.hg.1 | ENST00000556362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228087.hg.1 | ENST00000622580.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228087.hg.1 | NM_003281.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228087.hg.1 | TC0100016914.hg.1 | Clariom D TC |

TNNI1
Genomic Map