Assay Details
| Gene Symbol: ZNF300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052860.2 | NP_443092.1 | 2 - 3 | 392 | 90 | |
| GenBank mRNA | AF395541.1 | - | 2 - 3 | 238 | 90 | |
| AK096465.1 | - | 2 - 3 | 389 | 90 | ||
| BC117248.1 | - | 2 - 3 | 141 | 40125799 | 90 | |
| DA497503.1 | - | 2 - 3 | 393 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001944.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF395541.1,AK096465.1,BC117248.1,NM_052860.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05026979.hg.1 | ENST00000274599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026979.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026979.hg.1 | ENST00000427179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026979.hg.1 | ENST00000446148.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026979.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026979.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05026979.hg.1 | TC05001944.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000418587.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_001172832.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012491.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187061.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187061.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187061.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187061.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187061.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187061.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187061.hg.1 | ENST00000274599.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187061.hg.1 | ENST00000394226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187061.hg.1 | ENST00000427179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187061.hg.1 | ENST00000446148.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500187061.hg.1 | NM_001172831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187061.hg.1 | NM_052860.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500187061.hg.1 | TC0500012491.hg.1 | Clariom D TC |

ZNF300
Genomic Map