Assay Details
| Gene Symbol: KDR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002253.2 | NP_002244.1 | 26 - 27 | 3808 | 103 | |
| GenBank mRNA | AB209901.1 | - | 26 - 27 | 3793 | 103 | |
| AF035121.1 | - | 26 - 27 | 3809 | 103 | ||
| AF063658.1 | - | 26 - 27 | 3506 | 103 | ||
| BC131822.1 | - | 26 - 27 | 3568 | 40129287 | 103 | |
| L04947.1 | - | 26 - 27 | 3500 | 103 | ||
| X61656.1 | - | 26 - 27 | 3500 | 103 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001208.hg.1
Set Membership: |
|

KDR
Genomic Map