Assay Details
| Gene Symbol: ARHGAP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018460.3 | NP_060930.3 | 12 - 13 | 1285 | 115 | |
| XM_011511479.2 | XP_011509781.1 | 13 - 14 | 1317 | 115 | ||
| XM_011511481.2 | XP_011509783.1 | 12 - 13 | 1220 | 115 | ||
| XM_011511482.2 | XP_011509784.1 | 12 - 13 | 1220 | 115 | ||
| XM_011511483.1 | XP_011509785.1 | 8 - 9 | 915 | 115 | ||
| XM_017004499.1 | XP_016859988.1 | 12 - 13 | 1246 | 115 | ||
| XM_017004500.1 | XP_016859989.1 | 11 - 12 | 1112 | 115 | ||
| GenBank mRNA | AF212222.1 | - | 12 - 13 | 1238 | 115 | |
| AF217507.1 | - | 12 - 13 | 1234 | 115 | ||
| AF217522.1 | - | 12 - 13 | 1239 | 115 | ||
| AK226014.1 | - | 12 - 13 | 1220 | 115 | ||
| AY219338.1 | - | 12 - 13 | 1223 | 115 | ||
| BC016701.2 | - | 12 - 13 | 1191 | 4108644 | 115 | |
| BC038976.1 | - | 12 - 13 | 1190 | 5553184 | 115 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000904.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02013336.hg.1 | ENST00000295095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013336.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013336.hg.1 | TC02000904.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000419455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000419455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000419455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000469117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000469117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000469117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000469117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000474474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000474474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000474474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000474474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000549060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000549060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000549060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200166673.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200166673.hg.1 | ENST00000549436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200166673.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200166673.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200166673.hg.1 | ENST00000549436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200166673.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200166673.hg.1 | TC0200009539.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000419455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000419455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000419455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000469117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000469117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000469117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000469117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000474474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000474474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000474474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000474474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548929.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548929.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548929.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548929.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000549060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000549060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000549060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |

ARHGAP15
Genomic Map