Assay Details
| Gene Symbol: LOC107985946 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017005480.1 | XP_016860969.1 | 1 | 954 | 56 | |
| GenBank mRNA | AK056055.1 | - | 1 | 647 | 56 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | R3H domain containing 1 | R3HDM1 | R3HDM | -16.5kb | Sense | Chr. 2 - 135531429 - 135725270 on NCBI Build GRCh38 |
| Downstream | lactase | LCT |
LAC, LPH, LPH1 |
45.5kb | Anti-sense | Chr. 2 - 135787840 - 135837200 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000415164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000415164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000415164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000415164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000415164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000415164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000415164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000415164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000426921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000426921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000426921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000426921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000426921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000426921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000426921.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000467065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000467065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000470687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000470687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000470687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000470687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000470687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000470687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000471246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000471246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000471246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000471246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000272638.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000415164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000415164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000415164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000415164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000415164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000415164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000415164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000415164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000416538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000426921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000426921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000426921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000426921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000426921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000426921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000426921.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000467065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000467065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000470687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000470687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000470687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000470687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000470687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000470687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000471246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000471246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000471246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000471246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | ENST00000490163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009447.hg.1 | NM_014607.3 | Refseq |

LOC107985946
Genomic Map