Assay Details
| Gene Symbol: KMT2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003482.3 | NP_003473.3 | 18 - 19 | 4738 | 66 | |
| XM_005269162.4 | XP_005269219.1 | 19 - 20 | 6017 | 66 | ||
| XM_006719614.3 | XP_006719677.1 | 19 - 20 | 6017 | 66 | ||
| XM_006719616.3 | XP_006719679.1 | 19 - 20 | 6014 | 66 | ||
| XM_011538770.2 | XP_011537072.1 | 19 - 20 | 6020 | 66 | ||
| XM_011538771.2 | XP_011537073.1 | 19 - 20 | 6017 | 66 | ||
| XM_011538772.2 | XP_011537074.1 | 19 - 20 | 6020 | 66 | ||
| XM_011538773.2 | XP_011537075.1 | 19 - 20 | 6017 | 66 | ||
| XM_011538774.2 | XP_011537076.1 | 19 - 20 | 6021 | 66 | ||
| XM_011538776.2 | XP_011537078.1 | 19 - 20 | 6020 | 66 | ||
| GenBank mRNA | AW997729.1 | - | 1 - 2 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12003277.hg.1
Set Membership: |
|

KMT2D
Genomic Map