Assay Details
| Gene Symbol: ZC3HAV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020119.3 | NP_064504.2 | 11 - 12 | 2708 | 86 | |
| XM_005250501.2 | XP_005250558.1 | 11 - 12 | 3075 | 86 | ||
| GenBank mRNA | AK023350.1 | - | 8 - 9 | 1081 | 86 | |
| AK292811.1 | - | 11 - 12 | 2376 | 86 | ||
| BC027462.1 | - | 3 - 4 | 295 | 86 | ||
| BX571742.1 | - | 11 - 12 | 2707 | 86 | ||
| BX647138.1 | - | 8 - 9 | 1439 | 86 | ||
| BX647974.1 | - | 11 - 12 | 2344 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001910.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
response to virus
positive regulation of type I interferon production
suppression by virus of host molecular function
negative regulation of viral genome replication
innate immune response
regulation of defense response to virus by host
defense response to virus
positive regulation of mRNA catabolic process
cellular response to exogenous dsRNA
cell-cell adhesion
NAD+ ADP-ribosyltransferase activity
protein binding
poly(A) RNA binding
metal ion binding
cadherin binding involved in cell-cell adhesion
response to virus
positive regulation of type I interferon production
suppression by virus of host molecular function
negative regulation of viral genome replication
innate immune response
regulation of defense response to virus by host
defense response to virus
positive regulation of mRNA catabolic process
cellular response to exogenous dsRNA
cell-cell adhesion
NAD+ ADP-ribosyltransferase activity
protein binding
poly(A) RNA binding
metal ion binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000460845.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000460845.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000460845.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000460845.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000460845.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000460845.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000460845.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029623.hg.1 | ENST00000242351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029623.hg.1 | ENST00000464606.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029623.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029623.hg.1 | TC07001910.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700191996.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700191996.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700191996.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700191996.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700191996.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700191996.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700191996.hg.1 | TC0700012761.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000242351.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000460845.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000460845.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000460845.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000460845.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000460845.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000460845.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000460845.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000464606.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | ENST00000471652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_020119.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_024625.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012761.hg.1 | NM_024625.3 | Refseq |

ZC3HAV1
Genomic Map