| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000417581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000417581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000417581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000417581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000417581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000421701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000421701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000421701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
ENST00000421701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014418.hg.1 |
ENST00000263063.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014418.hg.1 |
ENST00000358107.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014418.hg.1 |
ENST00000488290.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014418.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014418.hg.1 |
TC10001150.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014419.hg.1 |
ENST00000263063.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014419.hg.1 |
ENST00000358107.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014419.hg.1 |
ENST00000488290.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014419.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014419.hg.1 |
TC10001150.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000186834.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000186834.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000186834.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000186834.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000186834.hg.1 |
TC1000012538.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000417581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000417581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000417581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000417581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000417581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000421701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000421701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000421701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
ENST00000421701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012538.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000186835.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000186835.hg.1 |
ENST00000417581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000186835.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000186835.hg.1 |
ENST00000263063.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000186835.hg.1 |
ENST00000417581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000186835.hg.1 |
NM_018109.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000186835.hg.1 |
TC1000012538.hg.1 |
Clariom D TC |