| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11022313.hg.1 |
ENST00000323646.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11022313.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11022313.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11022313.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11022313.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11022313.hg.1 |
TC11001893.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000540943.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000540943.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000540943.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000544269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000544269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000544269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
ENST00000544269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100184817.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184817.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184817.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184817.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100184817.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100184817.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184817.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184817.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184817.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100184817.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100184817.hg.1 |
TC1100011126.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100184818.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184818.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184818.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184818.hg.1 |
ENST00000540943.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184818.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100184818.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100184818.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184818.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184818.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184818.hg.1 |
ENST00000540943.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184818.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100184818.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100184818.hg.1 |
TC1100011126.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100184819.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184819.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184819.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184819.hg.1 |
ENST00000540943.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184819.hg.1 |
ENST00000544269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184819.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100184819.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100184819.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184819.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184819.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184819.hg.1 |
ENST00000540943.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184819.hg.1 |
ENST00000544269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100184819.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100184819.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100184819.hg.1 |
TC1100011126.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000323646.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000394613.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000415826.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000540943.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000540943.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000540943.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000544269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000544269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000544269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
ENST00000544269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_001128203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011126.hg.1 |
NM_007069.3 |
Refseq |