Assay Details
| Gene Symbol: ARID3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005259513.4 | XP_005259570.1 | 9 | 5381 | 132 | |
| XM_005259514.4 | XP_005259571.1 | 9 | 5064 | 132 | ||
| XM_017026445.1 | XP_016881934.1 | 9 | 5090 | 132 | ||
| XM_017026446.1 | XP_016881935.1 | 7 | 4133 | 132 | ||
| XM_017026447.1 | XP_016881936.1 | 7 | 3973 | 132 | ||
| GenBank mRNA | BC010544.2 | - | 1 | 513 | 132 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | KISS1 receptor | KISS1R |
AXOR12, CPPB1, GPR54, HH8, HOT7T175, KISS-1R |
-50.9kb | Sense | Chr. 19 - 916693 - 921015 on NCBI Build GRCh38 |
| Downstream | WD repeat domain 18 | WDR18 |
Ipi3, R32184_1 |
8.4kb | Sense | Chr. 19 - 982980 - 994570 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ARID3A
Genomic Map