Assay Details
| Gene Symbol: SLC12A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195483.1 | NP_001182412.1 | 11 - 12 | 1971 | 97 | |
| NM_024628.5 | NP_078904.3 | 12 - 13 | 2032 | 97 | ||
| GenBank mRNA | AF345197.1 | - | 11 - 12 | 1921 | 97 | |
| AF390442.1 | - | 11 - 12 | 1972 | 97 | ||
| AK026841.1 | - | 8 - 9 | 1368 | 97 | ||
| AK123001.1 | - | 12 - 13 | 1986 | 97 | ||
| AK308484.1 | - | 7 - 8 | 1289 | 97 | ||
| BC020506.1 | - | 4 - 5 | 934 | 97 | ||
| BC126158.1 | - | 11 - 12 | 1942 | 8991874 | 97 | |
| BC126160.1 | - | 11 - 12 | 1942 | 8991876 | 97 | |
| BX538023.1 | - | 6 - 7 | 1355 | 97 | ||
| CR457363.1 | - | 3 - 4 | 703 | 97 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03003391.hg.1
Set Membership: |
|

SLC12A8
Genomic Map