Assay Details
| Gene Symbol: SAMM50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015380.4 | NP_056195.3 | 4 - 5 | 515 | 118 | |
| GenBank mRNA | AF151809.1 | - | 4 - 5 | 427 | 118 | |
| AK001087.1 | - | 4 - 5 | 118 | |||
| AK222657.1 | - | 4 - 5 | 389 | 118 | ||
| AK312773.1 | - | 4 - 5 | 350 | 118 | ||
| AY189688.1 | - | 4 - 5 | 430 | 118 | ||
| BC007830.1 | - | 4 - 5 | 422 | 3635175 | 118 | |
| BC011681.2 | - | 4 - 5 | 475 | 3010765 | 118 | |
| BC015200.1 | - | 4 - 5 | 365 | 4333051 | 118 | |
| CR456483.1 | - | 4 - 5 | 327 | 118 | ||
| DB030637.1 | - | 4 - 5 | 515 | 118 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22001478.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000465768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000465768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000465768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000493621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000493621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000493621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000494795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000494795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000494795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000494795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000494795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | ENST00000494795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007023.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007023.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007023.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007023.hg.1 | TC22001478.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000465768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000465768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000465768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000474323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000493621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000493621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000493621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000494795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000494795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000494795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000494795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000494795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | ENST00000494795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009282.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169227.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169227.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169227.hg.1 | NM_015380.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169227.hg.1 | TC2200009282.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169227.hg.1 | ENST00000350028.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169227.hg.1 | ENST00000493161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169227.hg.1 | NM_015380.4 | Refseq |

SAMM50
Genomic Map