| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22007026.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22007026.hg.1 |
ENST00000396202.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22007026.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22007026.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22007026.hg.1 |
TC22001478.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000465768.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000465768.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000465768.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493621.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493621.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000493621.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000465768.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000465768.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000465768.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000474323.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493621.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493621.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000493621.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
ENST00000494795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009282.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200169229.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200169229.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200169229.hg.1 |
NM_015380.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200169229.hg.1 |
TC2200009282.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200169229.hg.1 |
ENST00000350028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200169229.hg.1 |
ENST00000493161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200169229.hg.1 |
NM_015380.4 |
Refseq |