| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011180.hg.1 |
ENST00000304877.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011180.hg.1 |
ENST00000392484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011180.hg.1 |
ENST00000527711.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011180.hg.1 |
ENST00000528193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011180.hg.1 |
ENST00000530868.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011180.hg.1 |
ENST00000532429.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011180.hg.1 |
ENST00000534000.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011180.hg.1 |
NM_001003395.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011180.hg.1 |
NM_003287.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011180.hg.1 |
TC06000958.hg.1 |
HTA TC |