| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023504.hg.1 |
ENST00000246801.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023504.hg.1 |
ENST00000358830.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023504.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023504.hg.1 |
TC19001723.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000599325.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000599325.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000599325.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000599325.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900193515.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900193515.hg.1 |
ENST00000358830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900193515.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900193515.hg.1 |
TC1900011160.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900193515.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900193515.hg.1 |
ENST00000358830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900193515.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000599325.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000599325.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000599325.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000599325.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |