| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025912.hg.1 |
ENST00000339697.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025912.hg.1 |
ENST00000368454.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025912.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025912.hg.1 |
TC06002071.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025911.hg.1 |
ENST00000339697.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025911.hg.1 |
ENST00000368454.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025911.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025911.hg.1 |
TC06002071.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025910.hg.1 |
ENST00000339697.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025910.hg.1 |
ENST00000368454.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025910.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06025910.hg.1 |
TC06002071.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600190071.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190071.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600190071.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190071.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600190071.hg.1 |
TC0600013054.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600190073.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190073.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600190073.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190073.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600190073.hg.1 |
TC0600013054.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600190072.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190072.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600190072.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190072.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600190072.hg.1 |
TC0600013054.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600190074.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190074.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600190074.hg.1 |
ENST00000339697.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190074.hg.1 |
NM_020755.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600190074.hg.1 |
TC0600013054.hg.1 |
Clariom D TC |