Assay Details
| Gene Symbol: SLC18A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135691.2 | NP_001129163.1 | 11 - 12 | 1488 | 94 | |
| NM_001142325.1 | NP_001135797.1 | 11 - 12 | 1488 | 94 | ||
| NM_003053.3 | NP_003044.1 | 10 - 11 | 1284 | 94 | ||
| XM_011544625.1 | XP_011542927.1 | 9 - 10 | 1133 | 94 | ||
| XM_011544627.1 | XP_011542929.1 | 10 - 11 | 1217 | 94 | ||
| GenBank mRNA | AK313720.1 | - | 10 - 11 | 1180 | 94 | |
| BC009387.2 | - | 10 - 11 | 1203 | 4127678 | 94 | |
| U39905.1 | - | 10 - 11 | 1284 | 94 | ||
More Information
Set Membership: |
|

SLC18A1
Genomic Map