Assay Details
| Gene Symbol: ZSCAN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027135.1 | 2 | 1608 | 67 | ||
| GenBank mRNA | AK096238.1 | - | 2 | 1576 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 135 | ZNF135 |
ZNF61, ZNF78L1, pHZ-17, pT3 |
-37.9kb | Anti-sense | Chr. 19 - 58059227 - 58069755 on NCBI Build GRCh38 |
| Downstream | zinc finger protein 329 | ZNF329 | 17.0kb | Sense | Chr. 19 - 58126328 - 58155169 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ZSCAN18
Genomic Map