Assay Details
| Gene Symbol: ADD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185054.1 | NP_001171983.1 | 14 - 15 | 2005 | 94 | |
| NM_001617.3 | NP_001608.1 | 14 - 15 | 2251 | 94 | ||
| XM_011532502.2 | XP_011530804.1 | 14 - 15 | 1992 | 94 | ||
| GenBank mRNA | AK291007.1 | - | 14 - 15 | 2206 | 94 | |
| BC008709.1 | - | 4 - 5 | 408 | 94 | ||
| BC010237.1 | - | 14 - 15 | 2185 | 94 | ||
| BC051882.1 | - | 14 - 15 | 2178 | 94 | ||
| BC056881.1 | - | 14 - 15 | 2034 | 94 | ||
| BC065525.1 | - | 14 - 15 | 2005 | 6142886 | 94 | |
More Information
Set Membership: |
|

ADD2
Genomic Map