Assay Details
| Gene Symbol: ADAMTS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030955.3 | NP_112217.2 | 13 - 14 | 2364 | 77 | |
| XM_017009905.1 | XP_016865394.1 | 13 - 14 | 2352 | 77 | ||
| XM_017009906.1 | XP_016865395.1 | 12 - 13 | 1753 | 77 | ||
| XM_017009907.1 | XP_016865396.1 | 5 - 6 | 645 | 77 | ||
| GenBank mRNA | AJ250725.1 | - | 13 - 14 | 2354 | 77 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001243.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
cell migration
proteoglycan catabolic process
negative regulation of chondrocyte differentiation
regulation of inflammatory response
proteolysis involved in cellular protein catabolic process
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to BMP stimulus
regulation of endothelial tube morphogenesis
negative regulation of hepatocyte growth factor receptor signaling pathway
negative regulation of cellular response to vascular endothelial growth factor stimulus
negative regulation of cellular response to hepatocyte growth factor stimulus
metalloendopeptidase activity
protein binding
zinc ion binding
cell-matrix adhesion
cell migration
proteoglycan catabolic process
negative regulation of chondrocyte differentiation
regulation of inflammatory response
proteolysis involved in cellular protein catabolic process
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to BMP stimulus
regulation of endothelial tube morphogenesis
negative regulation of hepatocyte growth factor receptor signaling pathway
negative regulation of cellular response to vascular endothelial growth factor stimulus
negative regulation of cellular response to hepatocyte growth factor stimulus
metalloendopeptidase activity
protein binding
zinc ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000352040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000504830.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000506952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000506952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000506952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000506952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000509762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000509762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000509762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000515401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000515401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000515401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000515401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000515401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | ENST00000515401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010453.hg.1 | NM_030955.3 | Refseq |

ADAMTS12
Genomic Map