| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009212.hg.1 |
ENST00000349129.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009212.hg.1 |
ENST00000484282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009212.hg.1 |
NM_001199942.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009212.hg.1 |
NM_015018.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009212.hg.1 |
TC06000768.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600160123.hg.1 |
ENST00000237163.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600160123.hg.1 |
ENST00000349129.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600160123.hg.1 |
ENST00000369739.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600160123.hg.1 |
ENST00000484282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600160123.hg.1 |
NM_001199942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600160123.hg.1 |
NM_015018.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600160123.hg.1 |
ENST00000237163.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600160123.hg.1 |
ENST00000349129.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600160123.hg.1 |
ENST00000369739.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600160123.hg.1 |
ENST00000484282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600160123.hg.1 |
NM_001199942.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600160123.hg.1 |
NM_015018.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600160123.hg.1 |
TC0600008663.hg.1 |
Clariom D TC |