Assay Details
| Gene Symbol: DIMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014473.2 | NP_055288.1 | 2 - 3 | 279 | 87 | |
| XM_011543323.2 | XP_011541625.1 | 2 - 3 | 317 | 87 | ||
| GenBank mRNA | AF102147.1 | - | 2 - 3 | 228 | 87 | |
| AK292723.1 | - | 2 - 3 | 307 | 87 | ||
| AK299480.1 | - | 2 - 3 | 269 | 87 | ||
| BC002841.2 | - | 2 - 3 | 276 | 3638494 | 87 | |
| BC010874.2 | - | 2 - 3 | 240 | 3878818 | 87 | |
| BM749030.1 | - | 2 - 3 | 289 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001401.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011543323.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05019579.hg.1 | ENST00000199320.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05019579.hg.1 | ENST00000506390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05019579.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05019579.hg.1 | TC05001401.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0500175951.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175951.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175951.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175951.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175951.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175951.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175951.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175951.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175951.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175951.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175951.hg.1 | TC0500010894.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000199320.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000506390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000509182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | ENST00000514911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | NM_014473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010894.hg.1 | NM_014473.3 | Refseq |

DIMT1
Genomic Map