Assay Details
| Gene Symbol: CDH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317214.1 | NP_001304143.1 | 7 - 8 | 1924 | 68 | |
| NM_004361.3 | NP_004352.2 | 7 - 8 | 1924 | 68 | ||
| NM_033646.2 | NP_387450.1 | 7 - 8 | 1554 | 68 | ||
| XM_017025523.1 | XP_016881012.1 | 7 - 8 | 2240 | 68 | ||
| GenBank mRNA | AB035301.1 | - | 7 - 8 | 1655 | 68 | |
| AJ007611.1 | - | 7 - 8 | 1552 | 68 | ||
| BC036786.1 | - | 7 - 8 | 1924 | 5744842 | 68 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000235.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000581601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | ENST00000581601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002796.hg.1 | ENST00000323011.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002796.hg.1 | ENST00000397966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002796.hg.1 | ENST00000397968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002796.hg.1 | ENST00000536984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002796.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002796.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002796.hg.1 | TC18000235.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002797.hg.1 | ENST00000323011.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002797.hg.1 | ENST00000397966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002797.hg.1 | ENST00000397968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002797.hg.1 | ENST00000536984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002797.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002797.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002797.hg.1 | TC18000235.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1800152013.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152013.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152013.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152013.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800152013.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800152013.hg.1 | TC1800007570.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800152013.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152013.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152013.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152013.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800152013.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000323011.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000397968.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000536984.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000581601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | ENST00000581601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_004361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_033646.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007570.hg.1 | NM_033646.2 | Refseq |

CDH7
Genomic Map