Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1227 | LINC01227 | -54.7kb | Sense | Chr. 16 - 80567103 - 80572808 on NCBI Build GRCh38 | |
| Downstream | prostate cancer-associated 47 | PRCAT47 | 200.4kb | Sense | Chr. 16 - 80828735 - 80892596 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map