Assay Details
| Gene Symbol: HTT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002111.8 | NP_002102.4 | 40 - 41 | 5520 | 77 | |
| GenBank mRNA | AB016794.1 | - | 40 - 41 | 5520 | 77 | |
| AB209506.1 | - | 26 - 27 | 6532 | 77 | ||
| L12392.1 | - | 40 - 41 | 5690 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000044.hg.1
Set Membership: |
|

HTT
Genomic Map