Assay Details
| Gene Symbol: HLA-G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017010817.1 | XP_016866306.1 | 2 - 3 | 349 | 84 | |
| XM_017010818.1 | XP_016866307.1 | 2 - 3 | 349 | 84 | ||
| GenBank mRNA | M90684.1 | - | 2 - 3 | 340 | 84 | |
| M90685.1 | - | 2 - 3 | 340 | 84 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
antigen processing and presentation of peptide antigen via MHC class I
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-independent
positive regulation of tolerance induction
positive regulation of T cell tolerance induction
immune response-inhibiting cell surface receptor signaling pathway
cellular defense response
antigen processing and presentation
positive regulation of interleukin-12 production
negative regulation of T cell proliferation
positive regulation of regulatory T cell differentiation
regulation of immune response
negative regulation of immune response
interferon-gamma-mediated signaling pathway
type I interferon signaling pathway
negative regulation of dendritic cell differentiation
antigen binding
receptor binding
peptide antigen binding
protein homodimerization activity
antigen processing and presentation of peptide antigen via MHC class I
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-independent
positive regulation of tolerance induction
positive regulation of T cell tolerance induction
immune response-inhibiting cell surface receptor signaling pathway
cellular defense response
antigen processing and presentation
positive regulation of interleukin-12 production
negative regulation of T cell proliferation
positive regulation of regulatory T cell differentiation
regulation of immune response
negative regulation of immune response
interferon-gamma-mediated signaling pathway
type I interferon signaling pathway
negative regulation of dendritic cell differentiation
antigen binding
receptor binding
peptide antigen binding
protein homodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |

HLA-G
Genomic Map