Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 443 | LINC00443 | -269.8kb | Anti-sense | Chr. 13 - 106653880 - 106672180 on NCBI Build GRCh38 | |
| Downstream | family with sequence similarity 155 member A | FAM155A | 226.0kb | Sense | Chr. 13 - 107168531 - 107867255 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map