Assay Details
| Gene Symbol: KCNK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104210.1 | 2 - 3 | 1184 | 66 | ||
| XM_017013531.1 | XP_016869020.1 | 2 - 3 | 950 | 66 | ||
| GenBank mRNA | AF257080.1 | - | 2 - 3 | 1126 | 66 | |
| BC075079.2 | - | 2 - 3 | 1169 | 30915596 | 66 | |
| BC075080.2 | - | 2 - 3 | 1169 | 30915383 | 66 | |
| BC112063.1 | - | 2 - 3 | 1169 | 8327531 | 66 | |
| BC112065.1 | - | 2 - 3 | 1169 | 8327533 | 66 | |
More Information
Set Membership: |
|

KCNK9
Genomic Map