Assay Details
| Gene Symbol: PPP3CB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289968.1 | NP_001276897.1 | 4 - 5 | 590 | 128 | |
| GenBank mRNA | M29550.1 | - | 4 - 5 | 562 | 128 | |
More Information
Set Membership: |
|

PPP3CB
Genomic Map