Assay Details
| Gene Symbol: DOPEY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199942.1 | NP_001186871.1 | 25 - 26 | 5905 | 261 | |
| NM_015018.3 | NP_055833.2 | 25 - 26 | 5932 | 261 | ||
| XM_011535619.2 | XP_011533921.1 | 24 - 25 | 5897 | 261 | ||
| XM_017010559.1 | XP_016866048.1 | 25 - 26 | 5927 | 261 | ||
| XM_017010560.1 | XP_016866049.1 | 24 - 25 | 5855 | 261 | ||
| XM_017010561.1 | XP_016866050.1 | 25 - 26 | 5879 | 261 | ||
| XM_017010562.1 | XP_016866051.1 | 24 - 25 | 5828 | 261 | ||
| XM_017010563.1 | XP_016866052.1 | 25 - 26 | 5906 | 261 | ||
| XM_017010564.1 | XP_016866053.1 | 25 - 26 | 5906 | 261 | ||
| XM_017010565.1 | XP_016866054.1 | 25 - 26 | 5900 | 261 | ||
| XM_017010566.1 | XP_016866055.1 | 25 - 26 | 5900 | 261 | ||
| XM_017010567.1 | XP_016866056.1 | 24 - 25 | 5828 | 261 | ||
| XM_017010568.1 | XP_016866057.1 | 24 - 25 | 5897 | 261 | ||
| XM_017010569.1 | XP_016866058.1 | 23 - 24 | 5814 | 261 | ||
| XM_017010570.1 | XP_016866059.1 | 24 - 25 | 5828 | 261 | ||
| XM_017010571.1 | XP_016866060.1 | 25 - 26 | 5927 | 156 | ||
| XM_017010572.1 | XP_016866061.1 | 25 - 26 | 5927 | 261 | ||
| XM_017010573.1 | XP_016866062.1 | 25 - 26 | 5927 | 261 | ||
| XM_017010574.1 | XP_016866063.1 | 25 - 26 | 5927 | 261 | ||
| GenBank mRNA | AB029040.2 | - | 8 - 9 | 2953 | 261 | |
| AK027030.1 | - | 5 - 6 | 1699 | 261 | ||
| BC144544.1 | - | 25 - 26 | 5803 | 9053071 | 261 | |
| BC150619.1 | - | 25 - 26 | 5803 | 9056989 | 261 | |
| BX648667.1 | - | 25 - 26 | 5930 | 261 | ||
More Information
Set Membership: |
|

DOPEY1
Genomic Map