Assay Details
| Gene Symbol: PUS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031307.3 | NP_112597.3 | 2 - 3 | 474 | 73 | |
| XM_005271688.3 | XP_005271745.1 | 2 - 3 | 472 | 73 | ||
| GenBank mRNA | AF325689.1 | - | 1 - 2 | 406 | 73 | |
| AK055702.1 | - | 2 - 3 | 451 | 73 | ||
| AK314251.1 | - | 2 - 3 | 472 | 73 | ||
| BC004822.1 | - | 2 - 3 | 463 | 3451112 | 73 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002424.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000529801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000529801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000529801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000529801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029059.hg.1 | ENST00000227474.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029059.hg.1 | ENST00000530811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029059.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029059.hg.1 | TC11002424.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100197784.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197784.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197784.hg.1 | ENST00000534158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197784.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197784.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197784.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197784.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197784.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197784.hg.1 | ENST00000534158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197784.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197784.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197784.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197784.hg.1 | TC1100012717.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100197782.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197782.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197782.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197782.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197782.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197782.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197782.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197782.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197782.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197782.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197782.hg.1 | TC1100012717.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100197785.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197785.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197785.hg.1 | ENST00000534158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197785.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197785.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197785.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197785.hg.1 | ENST00000534158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197785.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197785.hg.1 | TC1100012717.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000529801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000529801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000529801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000529801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197783.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197783.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197783.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197783.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197783.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197783.hg.1 | ENST00000227474.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197783.hg.1 | ENST00000530811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197783.hg.1 | ENST00000613398.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197783.hg.1 | NM_001271985.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197783.hg.1 | NM_031307.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197783.hg.1 | TC1100012717.hg.1 | Clariom D TC |

PUS3
Genomic Map