Assay Details
| Gene Symbol: TNFSF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190942.1 | NP_001177871.1 | 1 - 2 | 257 | 97 | |
| NM_003810.3 | NP_003801.1 | 1 - 2 | 257 | 97 | ||
| NR_033994.1 | 1 - 2 | 257 | 97 | |||
| GenBank mRNA | AK296085.1 | - | 1 - 2 | 225 | 97 | |
| AK312742.1 | - | 1 - 2 | 225 | 97 | ||
| BC009795.1 | - | 1 - 2 | 154 | 97 | ||
| BC032722.1 | - | 1 - 2 | 209 | 5504670 | 97 | |
| BT019564.1 | - | 1 - 2 | 134 | 97 | ||
| CR456895.1 | - | 1 - 2 | 134 | 97 | ||
| DA041872.1 | - | 1 - 2 | 217 | 97 | ||
| DA969653.1 | - | 1 - 2 | 257 | 97 | ||
| DQ848564.1 | - | 1 - 2 | 134 | 97 | ||
| EU183231.1 | - | 1 - 2 | 134 | 97 | ||
| U37518.1 | - | 1 - 2 | 221 | 97 | ||
| U57059.1 | - | 1 - 2 | 224 | 97 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002006.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
immune response
signal transduction
cell surface receptor signaling pathway
cell-cell signaling
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of release of cytochrome c from mitochondria
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
positive regulation of extrinsic apoptotic signaling pathway
receptor binding
cytokine activity
tumor necrosis factor receptor binding
protein binding
metal ion binding
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
immune response
signal transduction
cell surface receptor signaling pathway
cell-cell signaling
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of release of cytochrome c from mitochondria
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
positive regulation of extrinsic apoptotic signaling pathway
receptor binding
cytokine activity
tumor necrosis factor receptor binding
protein binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03035851.hg.1 | ENST00000241261.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035851.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035851.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035851.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035851.hg.1 | TC03002006.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035844.hg.1 | ENST00000241261.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035844.hg.1 | ENST00000420541.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035844.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035844.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035844.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035844.hg.1 | TC03002006.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000420541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000420541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000420541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000420541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000420541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000420541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000420541.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000430881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000430881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000430881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000430881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000466777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000466777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000466777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000466777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000472804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000472804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000472804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000472804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000472804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000472804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000494851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000494851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000494851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000494851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000494851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000494851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | ENST00000494851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_001190943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_001190943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_001190943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_001190943.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198244.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198244.hg.1 | ENST00000420541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198244.hg.1 | ENST00000430881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198244.hg.1 | ENST00000472804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198244.hg.1 | ENST00000494851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198244.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198244.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198244.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198244.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198244.hg.1 | ENST00000420541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198244.hg.1 | ENST00000430881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198244.hg.1 | ENST00000472804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198244.hg.1 | ENST00000494851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198244.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198244.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198244.hg.1 | TC0300013146.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300198244.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000420541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000420541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000420541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000420541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000420541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000420541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000420541.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000430881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000430881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000430881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000430881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000466777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000466777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000466777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000466777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000472804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000472804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000472804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000472804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000472804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000472804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000494851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000494851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000494851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000494851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000494851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000494851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | ENST00000494851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_001190943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_001190943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_001190943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_001190943.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013146.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198252.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198252.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198252.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198252.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198252.hg.1 | ENST00000241261.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198252.hg.1 | NM_001190942.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198252.hg.1 | NM_003810.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198252.hg.1 | NR_033994.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198252.hg.1 | TC0300013146.hg.1 | Clariom D TC |

TNFSF10
Genomic Map