| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011174.hg.1 |
ENST00000283875.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011174.hg.1 |
NM_005513.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011174.hg.1 |
TC03000619.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011179.hg.1 |
ENST00000283875.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011179.hg.1 |
NM_005513.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011179.hg.1 |
TC03000619.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011177.hg.1 |
ENST00000283875.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011177.hg.1 |
NM_005513.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011177.hg.1 |
TC03000619.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011178.hg.1 |
ENST00000283875.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011178.hg.1 |
NM_005513.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011178.hg.1 |
TC03000619.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300160619.hg.1 |
ENST00000283875.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160619.hg.1 |
ENST00000469772.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160619.hg.1 |
ENST00000484715.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160619.hg.1 |
ENST00000492959.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160619.hg.1 |
ENST00000497393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160619.hg.1 |
NM_005513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160619.hg.1 |
ENST00000283875.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160619.hg.1 |
ENST00000469772.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160619.hg.1 |
ENST00000484715.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160619.hg.1 |
ENST00000492959.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160619.hg.1 |
ENST00000497393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160619.hg.1 |
NM_005513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160619.hg.1 |
TC0300008518.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300160624.hg.1 |
ENST00000283875.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160624.hg.1 |
ENST00000484715.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160624.hg.1 |
ENST00000492959.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160624.hg.1 |
ENST00000497393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160624.hg.1 |
NM_005513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160624.hg.1 |
ENST00000283875.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160624.hg.1 |
ENST00000484715.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160624.hg.1 |
ENST00000492959.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160624.hg.1 |
ENST00000497393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160624.hg.1 |
NM_005513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160624.hg.1 |
TC0300008518.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300160625.hg.1 |
ENST00000283875.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160625.hg.1 |
ENST00000492959.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160625.hg.1 |
ENST00000497393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160625.hg.1 |
NM_005513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160625.hg.1 |
ENST00000283875.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160625.hg.1 |
ENST00000492959.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160625.hg.1 |
ENST00000497393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160625.hg.1 |
NM_005513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160625.hg.1 |
TC0300008518.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300160623.hg.1 |
ENST00000283875.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160623.hg.1 |
ENST00000484715.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160623.hg.1 |
ENST00000492959.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160623.hg.1 |
ENST00000497393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160623.hg.1 |
NM_005513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160623.hg.1 |
ENST00000283875.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160623.hg.1 |
ENST00000484715.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160623.hg.1 |
ENST00000492959.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160623.hg.1 |
ENST00000497393.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160623.hg.1 |
NM_005513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160623.hg.1 |
TC0300008518.hg.1 |
Clariom D TC |