Assay Details
| Gene Symbol: PGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024419.4 | NP_077733.3 | 3 - 4 | 461 | 102 | |
| NR_111989.1 | 2 - 3 | 271 | 102 | |||
| XM_011525487.2 | XP_011523789.1 | 3 - 4 | 836 | 102 | ||
| XM_011525488.2 | XP_011523790.1 | 3 - 4 | 836 | 102 | ||
| XM_011525489.2 | XP_011523791.1 | 3 - 4 | 836 | 102 | ||
| XM_017025357.1 | XP_016880846.1 | 3 - 4 | 836 | 102 | ||
| XM_017025358.1 | XP_016880847.1 | 3 - 4 | 836 | 102 | ||
| XM_017025359.1 | XP_016880848.1 | 3 - 4 | 836 | 102 | ||
| XM_017025360.1 | XP_016880849.1 | 3 - 4 | 836 | 102 | ||
| XM_017025361.1 | XP_016880850.1 | 3 - 4 | 455 | 102 | ||
| XM_017025362.1 | XP_016880851.1 | 3 - 4 | 455 | 102 | ||
| XM_017025363.1 | XP_016880852.1 | 3 - 4 | 407 | 102 | ||
| XM_017025366.1 | XP_016880855.1 | 3 - 4 | 455 | 102 | ||
| GenBank mRNA | AK296882.1 | - | 3 - 4 | 435 | 102 | |
| AK307874.1 | - | 3 - 4 | 437 | 102 | ||
| AK316147.1 | - | 3 - 4 | 439 | 102 | ||
| BC008903.2 | - | 3 - 4 | 409 | 102 | ||
| BC015570.2 | - | 3 - 4 | 409 | 102 | ||
| BC025951.1 | - | 3 - 4 | 394 | 102 | ||
| BC108732.1 | - | 3 - 4 | 425 | 6504169 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

PGS1
Genomic Map