| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18000167.hg.1 |
ENST00000254528.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18000167.hg.1 |
ENST00000308080.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18000167.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18000167.hg.1 |
TC18000014.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000583776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000583776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000583776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000583776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000583776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
ENST00000583776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000583776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000583776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000583776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000583776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000583776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
ENST00000583776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006493.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800144499.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800144499.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800144499.hg.1 |
ENST00000583776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800144499.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800144499.hg.1 |
TC1800006493.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800144499.hg.1 |
NM_032048.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800144499.hg.1 |
ENST00000254528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800144499.hg.1 |
ENST00000308080.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800144499.hg.1 |
ENST00000583776.1 |
Ensembl |