Assay Details
| Gene Symbol: TTLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012263.4 | NP_036395.1 | 4 - 5 | 563 | 80 | |
| XM_011530108.2 | XP_011528410.1 | 5 - 6 | 590 | 80 | ||
| XM_017028751.1 | XP_016884240.1 | 5 - 6 | 565 | 80 | ||
| XM_017028752.1 | XP_016884241.1 | 4 - 5 | 476 | 80 | ||
| XM_017028753.1 | XP_016884242.1 | 5 - 6 | 665 | 80 | ||
| GenBank mRNA | AF104927.1 | - | 4 - 5 | 464 | 80 | |
| AK095996.1 | - | 4 - 5 | 539 | 80 | ||
| AK315658.1 | - | 4 - 5 | 488 | 80 | ||
| AL096883.1 | - | 4 - 5 | 483 | 80 | ||
| AL096886.1 | - | 3 - 4 | 411 | 80 | ||
| AL136687.1 | - | 4 - 5 | 563 | 80 | ||
| AL589867.1 | - | 2 - 3 | 322 | 80 | ||
| BC014968.1 | - | 4 - 5 | 550 | 4876418 | 80 | |
| CR456599.1 | - | 2 - 3 | 326 | 80 | ||
| DA247656.1 | - | 4 - 5 | 516 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000857.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22014487.hg.1 | ENST00000266254.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014487.hg.1 | ENST00000331018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014487.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014487.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014487.hg.1 | TC22000857.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014489.hg.1 | ENST00000266254.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014489.hg.1 | ENST00000331018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014489.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014489.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014489.hg.1 | TC22000857.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200164564.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164564.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164564.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164564.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164564.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200164564.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200164564.hg.1 | TC2200008898.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200164564.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164564.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164564.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164564.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164564.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200164564.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000266254.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000331018.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000439248.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | ENST00000440761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NM_012263.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NR_027779.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008898.hg.1 | NR_027779.1 | Refseq |

TTLL1
Genomic Map