Assay Details
| Gene Symbol: SMPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045617.1 | 5 - 6 | 613 | 148 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000915.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012353.hg.1 | ENST00000379494.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012353.hg.1 | ENST00000494525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012353.hg.1 | NM_014332.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012353.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012353.hg.1 | TC0X000915.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | ENST00000379494.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | ENST00000379494.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | ENST00000379494.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | ENST00000379494.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | ENST00000379494.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | ENST00000379494.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | ENST00000379494.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | ENST00000494525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | ENST00000494525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | ENST00000494525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | ENST00000494525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | ENST00000494525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | ENST00000494525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | NM_014332.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | NM_014332.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | NM_014332.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | NM_014332.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | NM_014332.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | NM_014332.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | NM_014332.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009227.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | ENST00000379494.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | ENST00000379494.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | ENST00000379494.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | ENST00000379494.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | ENST00000379494.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | ENST00000379494.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | ENST00000379494.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | ENST00000494525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | ENST00000494525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | ENST00000494525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | ENST00000494525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | ENST00000494525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | ENST00000494525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | NM_014332.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | NM_014332.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | NM_014332.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | NM_014332.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | NM_014332.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | NM_014332.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | NM_014332.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009227.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163429.hg.1 | ENST00000379494.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163429.hg.1 | ENST00000494525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163429.hg.1 | NM_014332.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163429.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163429.hg.1 | ENST00000379494.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163429.hg.1 | ENST00000494525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163429.hg.1 | NM_014332.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163429.hg.1 | NR_045617.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163429.hg.1 | TC0X00009227.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 141034 | - | - |

SMPX
Genomic Map