Assay Details
| Gene Symbol: LINC01213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110168.1 | 1 - 2 | 83 | |||
| GenBank mRNA | AW195433.1 | - | 2 - 3 | 387 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01213| Gene Symbol: LINC01213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110168.1 | 1 - 2 | 83 | |||
| GenBank mRNA | AW195433.1 | - | 2 - 3 | 387 | 83 | |
Set Membership: |
|