Assay Details
| Gene Symbol: LINC01094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038306.1 | 2 - 3 | 371 | 77 | ||
| GenBank mRNA | AK057213.1 | - | 2 - 3 | 148 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01094| Gene Symbol: LINC01094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038306.1 | 2 - 3 | 371 | 77 | ||
| GenBank mRNA | AK057213.1 | - | 2 - 3 | 148 | 77 | |
Set Membership: |
|