Assay Details
| Gene Symbol: GYG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184702.1 | NP_001171631.1 | 9 - 10 | 1431 | 67 | |
| XM_017029929.1 | XP_016885418.1 | 10 - 11 | 1634 | 67 | ||
| GenBank mRNA | U94363.1 | - | 9 - 10 | 1305 | 67 | |
More Information
Set Membership: |
|

GYG2
Genomic Map