Assay Details
| Gene Symbol: CERK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022766.5 | NP_073603.2 | 6 - 7 | 825 | 64 | |
| XM_017028909.1 | XP_016884398.1 | 6 - 7 | 622 | 64 | ||
| GenBank mRNA | AB051433.1 | - | 5 - 6 | 551 | 64 | |
| AB079066.1 | - | 6 - 7 | 841 | 64 | ||
| AJ457828.1 | - | 6 - 7 | 788 | 64 | ||
| AK291476.1 | - | 6 - 7 | 781 | 64 | ||
| BC067255.1 | - | 5 - 6 | 702 | 64 | ||
| BC126940.1 | - | 6 - 7 | 768 | 40084110 | 64 | |
| CR456404.1 | - | 6 - 7 | 728 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000890.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22014977.hg.1 | ENST00000216264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014977.hg.1 | ENST00000541677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014977.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014977.hg.1 | TC22000890.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000460254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000460254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000460254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000460254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165416.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165416.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165416.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165416.hg.1 | TC2200009028.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200165416.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165416.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165416.hg.1 | NM_022766.5 | Refseq |

CERK
Genomic Map