| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
ENST00000299853.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
ENST00000359210.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
ENST00000418581.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
ENST00000563282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
ENST00000564209.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
ENST00000564750.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
ENST00000565117.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
ENST00000615879.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
NM_001258033.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
NM_001258034.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
NM_001258035.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
NM_001258036.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
NM_018119.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
NR_047581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
TC1600007193.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
ENST00000299853.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
ENST00000359210.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
ENST00000418581.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
ENST00000563282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
ENST00000564209.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
ENST00000564750.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
ENST00000565117.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
ENST00000615879.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
NM_001258033.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
NM_001258034.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
NM_001258035.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
NM_001258036.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
NM_018119.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600151649.hg.1 |
NR_047581.1 |
Refseq |