Assay Details
| Gene Symbol: TUBGCP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286414.2 | NP_001273343.1 | 7 - 8 | 1023 | 69 | |
| NM_014444.4 | NP_055259.2 | 7 - 8 | 1023 | 69 | ||
| XM_011521454.2 | XP_011519756.1 | 7 - 8 | 1023 | 69 | ||
| XM_011521455.2 | XP_011519757.1 | 7 - 8 | 921 | 69 | ||
| XM_017022078.1 | XP_016877567.1 | 7 - 8 | 1023 | 69 | ||
| GenBank mRNA | AJ249677.1 | - | 7 - 8 | 729 | 69 | |
| AK001639.1 | - | 3 - 4 | 334 | 69 | ||
| AK026551.1 | - | 7 - 8 | 1010 | 69 | ||
| AK027703.1 | - | 7 - 8 | 981 | 69 | ||
| BC000966.2 | - | 5 - 6 | 483 | 3446803 | 69 | |
| BC009870.2 | - | 7 - 8 | 948 | 3940649 | 69 | |
| BC012801.2 | - | 7 - 8 | 967 | 2821891 | 69 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000328.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
microtubule nucleation
meiotic nuclear division
cytoplasmic microtubule organization
centrosome duplication
interphase microtubule nucleation by interphase microtubule organizing center
mitotic spindle assembly
structural constituent of cytoskeleton
protein binding
gamma-tubulin binding
microtubule minus-end binding
protein complex assembly
microtubule nucleation
meiotic nuclear division
cytoplasmic microtubule organization
centrosome duplication
interphase microtubule nucleation by interphase microtubule organizing center
mitotic spindle assembly
structural constituent of cytoskeleton
protein binding
gamma-tubulin binding
microtubule minus-end binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000260383.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000561691.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000562053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000562053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000562053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000562053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000563963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000564511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000565548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000565548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000565548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000565548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000566251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000566251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000566251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000566251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000566251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000570081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000570081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000570081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000570081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | ENST00000570081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_001286414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007057.hg.1 | NM_014444.4 | Refseq |

TUBGCP4
Genomic Map