Assay Details
| Gene Symbol: UBE2T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310326.1 | NP_001297255.1 | 1 - 2 | 72 | 70 | |
| NM_014176.3 | NP_054895.1 | 1 - 2 | 72 | 70 | ||
| GenBank mRNA | AF161499.1 | - | 1 - 2 | 62 | 70 | |
| BC004152.2 | - | 1 - 2 | 34 | 2821806 | 70 | |
| BC019284.1 | - | 1 - 2 | 34 | 2821806 | 70 | |
| BG776416.1 | - | 1 - 2 | 73 | 70 | ||
| BM460257.1 | - | 1 - 2 | 45 | 70 | ||
| BP244265.1 | - | 1 - 2 | 72 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003702.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF161499.1,BC004152.2,BC019284.1,NM_001310326.1,NM_014176.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
protein monoubiquitination
cellular response to DNA damage stimulus
protein K29-linked ubiquitination
interstrand cross-link repair
protein K27-linked ubiquitination
protein autoubiquitination
protein K63-linked ubiquitination
protein K48-linked ubiquitination
protein K11-linked ubiquitination
protein K6-linked ubiquitination
chromatin binding
ubiquitin-protein transferase activity
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity
DNA repair
protein monoubiquitination
cellular response to DNA damage stimulus
protein K29-linked ubiquitination
interstrand cross-link repair
protein K27-linked ubiquitination
protein autoubiquitination
protein K63-linked ubiquitination
protein K48-linked ubiquitination
protein K11-linked ubiquitination
protein K6-linked ubiquitination
chromatin binding
ubiquitin-protein transferase activity
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01056848.hg.1 | ENST00000367274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056848.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056848.hg.1 | TC01003702.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000460852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000460852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000460852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000460852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000460852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000460852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056846.hg.1 | ENST00000367274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056846.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056846.hg.1 | TC01003702.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056847.hg.1 | ENST00000367274.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056847.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01056847.hg.1 | TC01003702.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100228392.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228392.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228392.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228392.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228392.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228392.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228392.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228392.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228392.hg.1 | TC0100016952.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100228391.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228391.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228391.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228391.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228391.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228391.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228391.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228391.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228391.hg.1 | TC0100016952.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100228390.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228390.hg.1 | ENST00000460852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228390.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228390.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228390.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228390.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228390.hg.1 | ENST00000460852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228390.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228390.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228390.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228390.hg.1 | TC0100016952.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100228389.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228389.hg.1 | ENST00000460852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228389.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228389.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228389.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228389.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228389.hg.1 | ENST00000460852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228389.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100228389.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228389.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100228389.hg.1 | TC0100016952.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000367274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000460852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000460852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000460852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000460852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000460852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000460852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | ENST00000487227.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_001310326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016952.hg.1 | NM_014176.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 208303 | - | - |

UBE2T
Genomic Map