| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000460852.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000460852.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000460852.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000460852.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000460852.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000460852.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01056841.hg.1 |
ENST00000367274.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01056841.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01056841.hg.1 |
TC01003702.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000460852.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000460852.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000460852.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000460852.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000460852.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000460852.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016952.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228383.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228383.hg.1 |
ENST00000460852.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228383.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228383.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228383.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228383.hg.1 |
ENST00000367274.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228383.hg.1 |
ENST00000460852.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228383.hg.1 |
ENST00000487227.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100228383.hg.1 |
NM_001310326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228383.hg.1 |
NM_014176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100228383.hg.1 |
TC0100016952.hg.1 |
Clariom D TC |