| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022863.hg.1 |
ENST00000304527.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022863.hg.1 |
ENST00000409252.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022863.hg.1 |
ENST00000409598.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022863.hg.1 |
ENST00000424281.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022863.hg.1 |
ENST00000427414.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022863.hg.1 |
ENST00000455740.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022863.hg.1 |
ENST00000514152.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022863.hg.1 |
NM_001009555.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022863.hg.1 |
NM_001128923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022863.hg.1 |
NM_001128924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022863.hg.1 |
NM_001243349.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022863.hg.1 |
TC04001637.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
ENST00000304527.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
ENST00000409252.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
ENST00000409598.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
ENST00000427414.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
ENST00000478503.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
ENST00000514152.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
NM_001009555.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
NM_001128923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
NM_001128924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
NM_001243349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
ENST00000304527.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
ENST00000409252.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
ENST00000409598.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
ENST00000427414.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
ENST00000478503.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
ENST00000514152.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
NM_001009555.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
NM_001128923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
NM_001128924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
NM_001243349.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400191185.hg.1 |
TC0400012977.hg.1 |
Clariom D TC |