| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
ENST00000229304.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
ENST00000229304.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
ENST00000229304.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
ENST00000229304.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
ENST00000229304.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
ENST00000467171.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
ENST00000467171.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
ENST00000467171.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
ENST00000467171.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
NM_001304566.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
NM_001304566.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
NM_001304566.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
NM_001304566.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
NM_001304566.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
NM_001644.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
NM_001644.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
NM_001644.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
NM_001644.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
NM_001644.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
NM_005889.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
NM_005889.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
NM_005889.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009789.hg.1 |
NM_005889.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12015491.hg.1 |
NM_001644.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12015491.hg.1 |
TC12001163.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12015486.hg.1 |
ENST00000229304.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12015486.hg.1 |
NM_001644.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12015486.hg.1 |
TC12001163.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12015490.hg.1 |
ENST00000229304.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12015490.hg.1 |
NM_001644.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12015490.hg.1 |
TC12001163.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200173546.hg.1 |
ENST00000229304.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200173546.hg.1 |
NM_001304566.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200173546.hg.1 |
NM_001644.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200173546.hg.1 |
ENST00000229304.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200173546.hg.1 |
NM_001304566.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200173546.hg.1 |
NM_001644.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200173546.hg.1 |
TC1200009789.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
ENST00000229304.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
ENST00000229304.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
ENST00000229304.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
ENST00000229304.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
ENST00000229304.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
ENST00000467171.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
ENST00000467171.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
ENST00000467171.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
ENST00000467171.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
NM_001304566.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
NM_001304566.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
NM_001304566.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
NM_001304566.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
NM_001304566.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
NM_001644.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
NM_001644.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
NM_001644.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
NM_001644.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
NM_001644.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
NM_005889.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
NM_005889.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
NM_005889.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009789.hg.1 |
NM_005889.3 |
Refseq |