Assay Details
| Gene Symbol: PLXDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282736.1 | NP_001269665.1 | 8 - 9 | 1557 | 92 | |
| NM_032812.8 | NP_116201.7 | 9 - 10 | 1704 | 92 | ||
| XM_011519750.2 | XP_011518052.1 | 9 - 10 | 1932 | 92 | ||
| GenBank mRNA | AF378757.1 | - | 9 - 10 | 1076 | 92 | |
| AK304591.1 | - | 10 - 11 | 1588 | 92 | ||
| AY358486.1 | - | 9 - 10 | 1704 | 92 | ||
| BC012885.1 | - | 8 - 9 | 1163 | 3865073 | 92 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000145.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001689.hg.1 | ENST00000377238.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001689.hg.1 | ENST00000377242.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001689.hg.1 | ENST00000377252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001689.hg.1 | ENST00000536022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001689.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001689.hg.1 | TC10000145.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006937.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147529.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147529.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147529.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147529.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147529.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147529.hg.1 | ENST00000377238.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147529.hg.1 | ENST00000377242.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147529.hg.1 | ENST00000377252.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000147529.hg.1 | NM_001282736.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147529.hg.1 | NM_032812.8 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000147529.hg.1 | TC1000006937.hg.1 | Clariom D TC |

PLXDC2
Genomic Map