| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05008714.hg.1 |
ENST00000296871.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05008714.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05008714.hg.1 |
TC05000629.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008620.hg.1 |
ENST00000296871.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008620.hg.1 |
ENST00000296871.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008620.hg.1 |
ENST00000296871.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008620.hg.1 |
ENST00000296871.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008620.hg.1 |
ENST00000296871.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008620.hg.1 |
ENST00000296871.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008620.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008620.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008620.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008620.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008620.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008620.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05008713.hg.1 |
ENST00000296871.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05008713.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05008713.hg.1 |
TC05000629.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0500159111.hg.1 |
ENST00000296871.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500159111.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500159111.hg.1 |
ENST00000296871.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500159111.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500159111.hg.1 |
TC0500008620.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500008620.hg.1 |
ENST00000296871.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008620.hg.1 |
ENST00000296871.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008620.hg.1 |
ENST00000296871.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008620.hg.1 |
ENST00000296871.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008620.hg.1 |
ENST00000296871.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008620.hg.1 |
ENST00000296871.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008620.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008620.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008620.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008620.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008620.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008620.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500159112.hg.1 |
ENST00000296871.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500159112.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500159112.hg.1 |
ENST00000296871.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500159112.hg.1 |
NM_000758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500159112.hg.1 |
TC0500008620.hg.1 |
Clariom D TC |