Assay Details
| Gene Symbol: CCDC162P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC128118.1 | - | 3 - 4 | 420 | 134 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC162P| Gene Symbol: CCDC162P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC128118.1 | - | 3 - 4 | 420 | 134 | |
Set Membership: |
|